missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylcarbamoyl)benzeneboronic acid, 98%

Product Code. 15429907
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15429907 250 mg 250mg
15419907 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15429907 Supplier Thermo Scientific Alfa Aesar Supplier No. H53141.MD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 832695-88-2 | C8H10BNO3 | 178.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 832695-88-2
Molecular Formula C8H10BNO3
Molecular Weight (g/mol) 178.98
MDL Number MFCD04038918
InChI Key FYFFPNFUVMBPRZ-UHFFFAOYSA-N
Synonym 3-n-methylaminocarbonyl phenylboronic acid, 3-methylcarbamoyl phenyl boronic acid, 3-methylcarbamoyl benzeneboronic acid, 3-methylcarbamoyl phenylboronic acid, 3-n-methylaminocarbonyl benzeneboronic acid, 3-borono-n-methylbenzamide, boronic acid, 3-methylamino carbonyl phenyl, acmc-209prq, ksc496e8l
PubChem CID 2773524
IUPAC Name [3-(methylcarbamoyl)phenyl]boronic acid
SMILES CNC(=O)C1=CC=CC(=C1)B(O)O

Specifications

Melting Point 114°C to 124°C
Quantity 250 mg
Formula Weight 178.98
Percent Purity 98%
Chemical Name or Material 3-(Methylcarbamoyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.