missing translation for 'onlineSavingsMsg'
Learn More

3-Methylcinnolin-5-amine, 97%, Thermo Scientific™

Product Code. 10734013
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10734013 1 g Amber Glass Bottle 1g
10110652 250 mg Amber Glass Bottle 250mg
10505374 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10734013 Supplier Thermo Scientific Maybridge Supplier No. RJC02157DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 300690-74-8
Molecular Formula C9H9N3
Molecular Weight (g/mol) 159.19
MDL Number MFCD00466524
InChI Key HOJQDQQMHMVETF-UHFFFAOYSA-N
PubChem CID 5310645
IUPAC Name 3-methylcinnolin-5-amine
SMILES CC1=NN=C2C=CC=C(N)C2=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 3-methyl-cinnolin-5-ylamine, 5-cinnolinamine, 3-methyl, 3-methyl-5-cinnolinamine, 3-methylcinnoline-5-ylamine, maybridge1_006688, 5-cinnolinamine,3-methyl, cambridge id 5256404, 3-methylcinnolin-5-yl amine, 5-amino-3-methyl-1,2-benzodiazine
Formula Weight 159.19
Percent Purity 95%
Chemical Name or Material 3-Methylcinnolin-5-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.