missing translation for 'onlineSavingsMsg'
Learn More

3-Methylglutaric anhydride, 97%

Product Code. 10222462 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10222462 5 g Glass bottle 5g
10562711 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10222462 Supplier Thermo Scientific Acros Supplier No. 126870050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4166-53-4 | C6H8O3 | 128.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4166-53-4
Molecular Formula C6H8O3
Molecular Weight (g/mol) 128.13
MDL Number MFCD00006683
InChI Key MGICRVTUCPFQQZ-UHFFFAOYSA-N
Synonym 3-methylglutaric anhydride, 4-methyldihydro-2h-pyran-2,6 3h-dione, 3-methylglutaricanhydride, .beta.-methylglutaric anhydride, acmc-1ago6, 2h-pyran-2,6 3h-dione, dihydro-4-methyl, 3-methyl glutaric anhydride, beta-methylglutaric anhydride, 4-methyltetrahydropyran-2,6-dione
PubChem CID 77828
IUPAC Name 4-methyloxane-2,6-dione
SMILES CC1CC(=O)OC(=O)C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 40°C to 45°C
Color White
Boiling Point 180°C to 182°C (25.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Beilstein 17, 415
Solubility Information Solubility in water: may decompose
Formula Weight 128.13
Percent Purity 97%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 3-Methylglutaric anhydride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methylglutaric anhydride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.