missing translation for 'onlineSavingsMsg'
Learn More

3-Methylglutaric Anhydride, TRC

Product Code. 30478426
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478426 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478426 Supplier Toronto Research Chemicals Supplier No. TRCM31155510G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Methylglutaric Anhydride
CAS 4166-53-4
Molecular Formula C31 H32 N2 O8
Quantity 10 g
Synonym Dihydro-4-methyl-2H-pyran-2,6(3H)-dione, 3-Methylglutaric Anhydride, 4-Methyltetrahydropyran-2,6-dione, beta-Methylglutaric Anhydride, beta-Methylglutaryl Anhydride
InChI Formula InChI=1S/C31H32N2O8/c1-37-23-14-12-21(13-15-23)31(20-8-5-4-6-9-20,22-10-7-11-24(18-22)38-2)40-19-25-27(35)28(39-3)29(41-25)33-17-16-26(34)32-30(33)36/h4-18,25,27-29,35H,19H2,1-3H3,(H,32,34,36)/t25-,27?,28?,29-,31?/m1/s1
SMILES COC1C(O)[C@@H](COC(c2ccccc2)(c3ccc(OC)cc3)c4cccc(OC)c4)O[C@H]1N5C=CC(=O)NC5=O
IUPAC Name 1-[(2R,5R)-4-hydroxy-3-methoxy-5-[[(3-methoxyphenyl)-(4-methoxyphenyl)-phenylmethoxy]methyl]oxolan-2-yl]pyrimidine-2,4-dione
Molecular Weight (g/mol) 560.594
Formula Weight 560.216
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.