missing translation for 'onlineSavingsMsg'
Learn More

3-Methylindole, 99%

Product Code. 11326706
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11326706

Brand: Thermo Scientific Alfa Aesar L03890.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 83-34-1 | C9H9N | 131.178 g/mol

3-Methylindole is often used as a component of commercial fragrances and perfumes.It helps fluorescent guanosine analogue, which in a dimethoxytrityl, phosphoramidite protected form, can be site-specifically inserted into oligonucleotides through a 3?5?phosphodiester linkage using an automated DNA synthesizer

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylindole is often used as a component of commercial fragrances and perfumes.It helps fluorescent guanosine analogue, which in a dimethoxytrityl, phosphoramidite protected form, can be site-specifically inserted into oligonucleotides through a 3?5?phosphodiester linkage using an automated DNA synthesizer

Solubility
Soluble in water, Ether, Alcohols, Benzene, Acetone, Chloroform.

Notes
Light Sensitive. Store in dark away from oxidizing agents. Incompatible with light, strong acids, acid ahydrides, acid chlorides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-34-1
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00005627
InChI Key ZFRKQXVRDFCRJG-UHFFFAOYSA-N
Synonym 3-methylindole, skatole, scatole, skatol, 1h-indole, 3-methyl, beta-methylindole, indole, 3-methyl, 3-mi, 3-methyl-4,5-benzopyrrole, 3-methyl indole
PubChem CID 6736
ChEBI CHEBI:9171
IUPAC Name 3-methyl-1H-indole
SMILES CC1=CNC2=CC=CC=C12

Specifications

Melting Point 94°C to 97°C
Boiling Point 265°C to 268°C
Flash Point 132°C (269°F)
Odor Stench
Quantity 25 g
Beilstein 111296
Sensitivity Light sensitive
Merck Index 14,8560
Solubility Information Soluble in water,Ether,Alcohols,Benzene,Acetone,Chloroform.
Formula Weight 131.18
Percent Purity 99%
Chemical Name or Material 3-Methylindole
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.