missing translation for 'onlineSavingsMsg'
Learn More

(3-Methylisoxazol-5-yl)methanol, 97%, Thermo Scientific™

Product Code. 10215484
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10215484 250 mg Amber Glass Bottle 250mg
10644215 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10215484 Supplier Thermo Scientific Maybridge Supplier No. CC27009CB
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 14716-89-3
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.116
MDL Number MFCD09859210
InChI Key OFSDWHRZVRCPBR-UHFFFAOYSA-N
Synonym 3-methylisoxazol-5-yl methanol, 3-methyl-1,2-oxazol-5-yl methanol, 3-methyl-5-isoxazolemethanol, 5-isoxazolemethanol,3-methyl, 3-methylisoxazol-5-yl methan-1-ol, 3-methylisoxazole-5-methanol, 5-isoxazolemethanol, 3-methyl, 3-methyl-5-isoxazolyl methanol, 5-hydroxymethyl-3-methylisoxazole, 5-hydroxymethyl-3-methyl-isoxazole
PubChem CID 10820493
IUPAC Name (3-methyl-1,2-oxazol-5-yl)methanol
SMILES CC1=NOC(=C1)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 113.12
Percent Purity 97%
Chemical Name or Material (3-Methylisoxazol-5-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.