missing translation for 'onlineSavingsMsg'
Learn More

3-Methylisoxazole-4-carboxylic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11372977
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11372977 1 g 1g
11382977 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372977 Supplier Thermo Scientific Alfa Aesar Supplier No. L16307.03
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17153-20-7 | Molecular Formula: C5H5NO3 | Molecular Weight: 127.099 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17153-20-7
Molecular Formula C5H5NO3
Molecular Weight (g/mol) 127.099
MDL Number MFCD01318161
InChI Key YBLSBWHFPXDRHC-UHFFFAOYSA-N
Synonym 3-methylisoxazole-4-carboxylic acid, 3-methyl-4-isoxazolecarboxylic acid, 3-methyl-4-isoxazolecarboxylicacid, 3-methyl-isoxazole-4-carboxylic acid, 4-isoxazolecarboxylic acid, 3-methyl, 3-methylisoxazole-4-carboxylicacid, pubchem10465, acmc-1c78i, 3-methylisoxazole-s-carboxylic acid, 4-isoxazolecarboxylicacid,3-methyl
PubChem CID 4589692
IUPAC Name 3-methyl-1,2-oxazole-4-carboxylic acid
SMILES CC1=NOC=C1C(=O)O

Specifications

Melting Point 185°C to 187°C
Quantity 1 g
Beilstein 114157
Formula Weight 127.1
Percent Purity ≥98%
Chemical Name or Material 3-Methylisoxazole-4-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.