missing translation for 'onlineSavingsMsg'
Learn More

3-Methylisoxazole-5-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 15710889 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15710889 1 g 1g
15289806 5 g 5g
15299806 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15710889 Supplier Thermo Scientific Acros Supplier No. 452660010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4857-42-5
Synonym 5-carboxy-3-methylisoxazole, 3-methylisoxazole-5-carboxylic acid, 3-methyl-5-isoxazolecarboxylic acid, 3-methyl-5-carboxyisoxazole, 5-isoxazolecarboxylicacid, 3-methyl, 5-isoxazolecarboxylic acid, 3-methyl, 3-methylisoxazole-5-carboxylicacid, aronis24519, 3-methyl-isoxazole-5-carboxylic acid, acmc-1aruu
PubChem CID 853085
IUPAC Name 3-methyl-1,2-oxazole-5-carboxylic acid

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C5H4NO3
Quantity 1 g
InChI Key HXIYCKAAQPHZBM-UHFFFAOYSA-M
SMILES CC1=NOC(=C1)C([O-])=O
Molecular Weight (g/mol) 126.09
Formula Weight 127.1
Percent Purity 95%
Chemical Name or Material 3-Methylisoxazole-5-carboxylic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.