missing translation for 'onlineSavingsMsg'
Learn More

3-Methylisoxazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10474815
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10474815 1 g Amber Glass Bottle 1g
10432494 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10474815 Supplier Thermo Scientific Maybridge Supplier No. CC27001DA
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 4857-42-5
Molecular Formula C5H5NO3
Molecular Weight (g/mol) 127.099
MDL Number MFCD00464222
InChI Key HXIYCKAAQPHZBM-UHFFFAOYSA-N
Synonym 3-methylisoxazole-5-carboxylic acid, 3-methyl-5-isoxazolecarboxylic acid, 5-isoxazolecarboxylic acid, 3-methyl, 3-methyl-isoxazole-5-carboxylic acid, 5-carboxy-3-methylisoxazole, 5-isoxazolecarboxylicacid, 3-methyl, 3-methylisoxazole-5-carboxylicacid, acmc-1aruu, 3-methyl-5-carboxyisoxazole, aronis24519
PubChem CID 853085
IUPAC Name 3-methyl-1,2-oxazole-5-carboxylic acid
SMILES CC1=NOC(=C1)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 127.1
Percent Purity 97%
Chemical Name or Material 3-Methylisoxazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.