missing translation for 'onlineSavingsMsg'
Learn More

3-Methylpentane, 99+%

Product Code. 10453825 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10453825 50 mL Glass bottle 50mL
10356960 250 mL Glass bottle 250mL
10329033 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10453825 Supplier Thermo Scientific Acros Supplier No. 127350500
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 96-14-0 | C6H14 | 86.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-14-0
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD00009342
InChI Key PFEOZHBOMNWTJB-UHFFFAOYSA-N
Synonym pentane, 3-methyl, unii-xd8o3ml76t, 3-methyl-pentane, xd8o3ml76t, 3-methylpentane, analytical standard, diethylmethylmethane, 3-methylpentyl, 3-methyl pentane, 3-methylpentan-3-yl, acmc-209s6l
PubChem CID 7282
IUPAC Name 3-methylpentane
SMILES CCC(C)CC

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -118.0°C
Color Colorless
Density 0.6600g/mL
Boiling Point 64.0°C
Flash Point −32°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula C2H5CH(CH3)C2H5
Refractive Index 1.3755 to 1.3775
Quantity 50 mL
Beilstein 01, 149
Specific Gravity 0.66
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, slightly soluble in ether
Formula Weight 86.18
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 3-Methylpentane, 99+%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Methylpentane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.