missing translation for 'onlineSavingsMsg'
Learn More

3-Methylpentane, TRC

Product Code. 30008044
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008044 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 30008044 Supplier Toronto Research Chemicals Supplier No. TRCM2263901G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 96-14-0
Molecular Formula C6 H14
Quantity 1 g
Synonym Pentane, 3-methyl-, 3-Methylpentane, NSC 66497, UN 1208
InChI Formula InChI=1S/C6H14/c1-4-6(3)5-2/h6H,4-5H2,1-3H3
SMILES CCC(C)CC
IUPAC Name 3-methylpentane
Molecular Weight (g/mol) 86.18
Formula Weight 86.1096
Chemical Name or Material 3-Methylpentane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.