missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-3-Methylpentanoic-d11 Acid, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 3-Methylpentanoic Acid-D11 |
| Molecular Formula | C6 D11 H O2 |
| Quantity | 0.25 g |
| Synonym | (±)-3-Methylpentanoic-d11 Acid, 3-Methylpentanoic Acid-D11, (±)-3-Methylvaleric Acid-d11 |
| InChI Formula | InChI=1 S/C6H12O2/c1-3-5(2)4-6(7)8/h5H,3-4H2,1-2H3,(H,7,8)/i1D3,2D3,3D2,4D2,5 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C(=O)O |
| IUPAC Name | 2,2,3,4,4,5,5,5-octadeuterio-3-(trideuteriomethyl)pentanoic acid |
| Molecular Weight (g/mol) | 127.2261 |
| Formula Weight | 127.1528 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction