missing translation for 'onlineSavingsMsg'
Learn More

(3-Methylphenyl)methanethiol, 95%, Thermo Scientific™

Product Code. 10565954
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10565954 25 g 25g
10496482 1 g 1g
10531703 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10565954 Supplier Thermo Scientific Maybridge Supplier No. TL00469EB
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 25697-56-7
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.228
MDL Number MFCD00022075
InChI Key PNAZUQUHTIOEHF-UHFFFAOYSA-N
PubChem CID 2801207
IUPAC Name (3-methylphenyl)methanethiol
SMILES CC1=CC(=CC=C1)CS

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Linear Formula CH3C6H4CH2SH
Quantity 25 g
Synonym 3-methylphenyl methanethiol, 3-methylbenzyl mercaptan, m-methylbenzyl mercaptan, 3-methylbenzenemethanethiol, m-methylbenzylmercaptan, acmc-1cq7k, 3-methyl-benzenemethanethiol, benzenemethanethiol,3-methyl, benzenemethanethiol, 3-methyl, 3-methylphenyl methane-1-thiol
Formula Weight 138.23
Percent Purity 95%
Chemical Name or Material (3-Methylphenyl)methanethiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.