missing translation for 'onlineSavingsMsg'
Learn More

3-Methylphthalic anhydride, 96%

Product Code. 10718481 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10718481 5 g Glass bottle 5g
10577552 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10718481 Supplier Thermo Scientific Acros Supplier No. 414780050
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 4792-30-7 | C9H6O3 | 162.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4792-30-7
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD00047316
InChI Key TWWAWPHAOPTQEU-UHFFFAOYSA-N
Synonym 3-methylphthalic anhydride, 4-methylisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 4-methyl, 3-methyl phthalic anhydride, methylphthalic anhydride, phthalic anhydride, 3-methyl, 2,3-toluenedicarboxylic anhydride, 1,3-isobenzofurandione, methyl, 4-methyl-1,3-dihydro-2-benzofuran-1,3-dione, acmc-1arrb
PubChem CID 98500
IUPAC Name 4-methyl-2-benzofuran-1,3-dione
SMILES CC1=CC=CC2=C1C(=O)OC2=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 114.0°C to 118.0°C
Color Beige to Brown-Yellow
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 5 g
Formula Weight 162.14
Percent Purity 96%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 3-Methylphthalic anhydride, 96%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Methylphthalic anhydride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.