missing translation for 'onlineSavingsMsg'
Learn More

3-Methylpyrazole, 99%

Product Code. 10254840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10254840 25 g Glass bottle 25g
10091470 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10254840 Supplier Thermo Scientific Acros Supplier No. 127560250
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1453-58-3 | C4H6N2 | 82.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1453-58-3
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00005240,MFCD08685900
InChI Key XKVUYEYANWFIJX-UHFFFAOYSA-N
PubChem CID 15073
IUPAC Name 5-methyl-1H-pyrazole
SMILES CC1=CC=NN1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 1.0200g/mL
Boiling Point 204°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.496
Quantity 25 g
Beilstein 23, 50
Specific Gravity 1.02
Synonym 3-methylpyrazole, 3-methyl-1h-pyrazole, 1h-pyrazole, 3-methyl, 5-methylpyrazole, 3 5-methylpyrazole, pyrazole, 5-methyl, 1h-pyrazole,5-methyl, 1h-pyrazole, 5-methyl, pyrazole, 3-methyl, unii-w7ku1rro6u
Solubility Information Solubility in water: miscible
Formula Weight 82.1
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Methylpyrazole, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Methylpyrazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.