missing translation for 'onlineSavingsMsg'
Learn More

3-Methylquinoxaline-2-carboxylic Acid, TRC

Product Code. 30136135
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136135 5 mg 5mg
30010295 50 mg 50mg
17913975 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136135 Supplier Toronto Research Chemicals Supplier No. TRCM3260605MG
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74003-63-7
Molecular Formula C10 H8 N2 O2
Molecular Weight (g/mol) 188.18
Synonym 2-Quinoxalinecarboxylic acid, 3-methyl- (7CI, 9CI, ACI), 3-Methyl-2-quinoxalinecarboxylic acid (ACI), MQCA
IUPAC Name 3-methylquinoxaline-2-carboxylic acid
SMILES Cc1nc2ccccc2nc1C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C10H8N2O2/c1-6-9(10(13)14)12-8-5-3-2-4-7(8)11-6/h2-5H,1H3,(H,13,14)
Formula Weight 188.0586
Percent Purity >95%
Chemical Name or Material 3-Methyl-quinoxaline-2-carboxylic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.