missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)propionaldehyde, 97%

Product Code. 11447846
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Product Code. Quantity unitSize
11447846 10 g 10g
11452487 50 g 50g
11462487 250 g 250g
3 options
This item is not returnable. View return policy

Product Code. 11447846

Brand: Thermo Scientific Alfa Aesar A14774.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3268-49-3 | C4H8OS | 104.167 g/mol

3-(Methylthio)propionaldehyde, is used as food flavour ,for making Baked food, spices, soft drinks, candy. It is also used as an intermediate. It has wide applications in organic synthesis as solvents as well as reagents. They are used in the synthesis of specific compound classes include agricultural chemical, property-enhancing additives, pharmacological drugs, chemical resistant polymers, detergents, and rubber antioxidants. Some examples of aliphatic and aromatic thioethers found in flavouring agents

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Methylthio)propionaldehyde, is used as food flavour ,for making Baked food, spices, soft drinks, candy. It is also used as an intermediate. It has wide applications in organic synthesis as solvents as well as reagents. They are used in the synthesis of specific compound classes include agricultural chemical, property-enhancing additives, pharmacological drugs, chemical resistant polymers, detergents, and rubber antioxidants. Some examples of aliphatic and aromatic thioethers found in flavouring agents

Solubility
It is readily soluble in alcohol solvents such as ethanol, propylene and glycol oil. It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C. Air sensitive. Handle and store under inert gas. Keep away from strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3268-49-3
Molecular Formula C4H8OS
Molecular Weight (g/mol) 104.167
MDL Number MFCD00007022
InChI Key CLUWOWRTHNNBBU-UHFFFAOYSA-N
Synonym methional, 3-methylthio propionaldehyde, 3-methylthio propanal, propanal, 3-methylthio, 4-thiapentanal, 3-methylsulfanyl propanal, 3-methylmercapto propionaldehyde, 3-methylthiopropanal, propionaldehyde, 3-methylthio, 3-methylmercaptopropyl aldehyde
PubChem CID 18635
ChEBI CHEBI:49017
IUPAC Name 3-methylsulfanylpropanal
SMILES CSCCC=O

Specifications

Melting Point -58°C
Density 1.052
Boiling Point 165°C to 166°C
Flash Point 64°C (147°F)
Odor Strong
Linear Formula CH3S(CH2)2CHO
Refractive Index 1.4825
Quantity 10 g
UN Number UN2785
Beilstein 1739289
Sensitivity Air sensitive
Solubility Information It is readily soluble in alcohol solvents such as ethanol,propylene and glycol oil. It is insoluble in water.
Formula Weight 104.17
Percent Purity 98%
Chemical Name or Material 3-(Methylthio)propionaldehyde
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-(Methylthio)propionaldehyde
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 4-Thiapentanal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.