missing translation for 'onlineSavingsMsg'
Learn More

3-Methylthiophene-2-carbonyl chloride, 98%, Thermo Scientific™

Product Code. 11364865
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11364865 10 g 10g
11354865 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11364865 Supplier Thermo Scientific Alfa Aesar Supplier No. L17989.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61341-26-2
Molecular Formula C6H5ClOS
Molecular Weight (g/mol) 160.62
MDL Number MFCD00130091
InChI Key AJSVPEVDFBYRCH-UHFFFAOYSA-N
Synonym 3-methyl-2-thiophenecarbonyl chloride, 3-methylthiophene-2-carbonylchloride, 3-methyl-2-thenoyl chloride, 2-3-methyl thiophene carboxylicacid chloride, pubchem5561, acmc-209mrl, 3-methylthenoyl chloride, 3-methyl-2-thenoylchloride, 3-methyl-2-thienoyl chloride, 3-methylthiophenecarbonylchloride
PubChem CID 2737605
IUPAC Name 3-methylthiophene-2-carbonyl chloride
SMILES CC1=C(SC=C1)C(Cl)=O

Specifications

Boiling Point 216°C to 220°C
Refractive Index 1.5855
Quantity 10 g
UN Number UN3265
Beilstein 116181
Sensitivity Moisture sensitive
Formula Weight 160.62
Percent Purity 98%
Chemical Name or Material 3-Methylthiophene-2-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.