missing translation for 'onlineSavingsMsg'
Learn More

3-Methylthiophene-2-methylamine, 96%

Product Code. 11354915
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354915 250 mg 250mg
11344915 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11354915 Supplier Thermo Scientific Alfa Aesar Supplier No. L18094.MD
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 104163-35-1 | C6H9NS | 127.205 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104163-35-1
Molecular Formula C6H9NS
Molecular Weight (g/mol) 127.205
MDL Number MFCD02093988
InChI Key SWZNXCABBUKIPZ-UHFFFAOYSA-N
PubChem CID 2798756
IUPAC Name (3-methylthiophen-2-yl)methanamine
SMILES CC1=C(SC=C1)CN

Specifications

Boiling Point 78°C (4 mmHg)
Refractive Index 1.5585
Quantity 250 mg
UN Number UN2735
Beilstein 108864
Sensitivity Air Sensitive
Synonym 3-methylthiophen-2-yl methanamine, 3-methyl-2-thienyl methylamine, 3-methylthiophene-2-methylamine, 2-thiophenemethanamine,3-methyl, 2-thiophenemethanamine, 3-methyl, acmc-20apo3, 3-metylthiophene-2-metylamine, 2-aminomethyl-3-methylthiophene, 3-methylthiophene-2-methanamine, 3-methylthien-2-yl methylamine
Formula Weight 127.21
Percent Purity 98%
Chemical Name or Material 3-Methylthiophene-2-methylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.