missing translation for 'onlineSavingsMsg'
Learn More

3-Methyluric Acid, TRC

Product Code. 30477572
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477572 25 mg 25mg
30481685 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30477572 Supplier Toronto Research Chemicals Supplier No. TRCM33819525MG
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 605-99-2
Molecular Formula C6 H6 N4 O3
Molecular Weight (g/mol) 182.14
Synonym N3-Methyluric Acid, 7,9-Dihydro-3-methyl-1H-purine-2,6,8(3H)-trione
IUPAC Name 3-methyl-7,9-dihydropurine-2,6,8-trione
SMILES CN1C(=O)NC(=O)C2=C1NC(=O)N2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C6H6N4O3/c1-10-3-2(7-5(12)8-3)4(11)9-6(10)13/h1H3,(H2,7,8,12)(H,9,11,13)
Formula Weight 182.044
Percent Purity >95%
Chemical Name or Material 3-Methyluric Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.