missing translation for 'onlineSavingsMsg'
Learn More

3-Methyluridine, MedChemExpress

Product Code. 16407487
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16407487 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16407487 Supplier Medchem Express Supplier No. HY113138
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 3-Methyluridine (N3-Methyluridine) is a modified RNA nucleoside. 3-Methyluridine is often presents as RNA modification which can be detected in 23S rRNA of archaea, 16S and 23S rRNA of eubacteria, and 18S, 25S, and 28S of eukaryotic ribosomal RNAs.

3-Methyluridine (N3-Methyluridine) is a modified RNA nucleoside. 3-Methyluridine is often presents as RNA modification which can be detected in 23S rRNA of archaea, 16S and 23S rRNA of eubacteria, and 18S, 25S, and 28S of eukaryotic ribosomal RNAs.

Specifications

Chemical Name or Material 3-Methyluridine
CAS 2140-69-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C10H14N2O6
Quantity 5 mg
Synonym N3-Methyluridine
Solubility Information DMSO : 160 mg/mL (619.60 mM; Need ultrasonic)
SMILES OC[C@@H]1[C@H]([C@H]([C@H](N2C(N(C)C(C=C2)=O)=O)O1)O)O
Molecular Weight (g/mol) 258.23
Formula Weight 258.23
Percent Purity 99.53%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.