missing translation for 'onlineSavingsMsg'
Learn More

3-Methylxanthine, 97%, Thermo Scientific™

Product Code. 13448458 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13448458 100 mg 100mg
13458458 250 mg 250mg
13438458 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13448458 Supplier Thermo Scientific Acros Supplier No. 206931000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1076-22-8
Molecular Formula C6H6N4O2
Molecular Weight (g/mol) 166.14
InChI Key GMSNIKWWOQHZGF-UHFFFAOYSA-N
Synonym 3-methylxanthine, 3-methyl-1h-purine-2,6 3h,7h-dione, 2,6-dihydroxy-3-methylpurine, xanthine, 3-methyl, 3 mx, 3-methyl xanthine, 1h-purine-2,6-dione, 3,7-dihydro-3-methyl, unii-ws6x982oec, 3-methyl-2,3,6,7-tetrahydro-1h-purine-2,6-dione, ccris 5817
PubChem CID 70639
ChEBI CHEBI:62207
IUPAC Name 3-methyl-7H-purine-2,6-dione
SMILES CN1C2=C(C(=O)NC1=O)NC=N2

Specifications

Melting Point >300.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Beilstein 26,453
Solubility Information Solubility in water: insoluble
Formula Weight 166.14
Percent Purity 97%
Chemical Name or Material 3-Methylxanthine
compliance-icons
Product Identifier
  • 3-Methylxanthine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.