missing translation for 'onlineSavingsMsg'
Learn More

3-Methylxanthine, 99.43%, For HPLC analysis, MP Biomedicals™

Product Code. 11452432 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452432 250 mg 250mg
11442432 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11452432 Supplier MP Biomedicals Supplier No. 0215168683
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1076-22-8
Molecular Formula C6H6N4O2
Molecular Weight (g/mol) 166.14
InChI Key GMSNIKWWOQHZGF-UHFFFAOYSA-N
Synonym 3-methylxanthine, 3-methyl-1h-purine-2,6 3h,7h-dione, 2,6-dihydroxy-3-methylpurine, xanthine, 3-methyl, 3 mx, 3-methyl xanthine, 1h-purine-2,6-dione, 3,7-dihydro-3-methyl, unii-ws6x982oec, 3-methyl-2,3,6,7-tetrahydro-1h-purine-2,6-dione, ccris 5817
PubChem CID 70639
ChEBI CHEBI:62207
IUPAC Name 3-methyl-7H-purine-2,6-dione
SMILES CN1C2=C(C(=O)NC1=O)NC=N2

Specifications

Color White
Quantity 250 mg
Formula Weight 166.1
Physical Form Powder
Chemical Name or Material 3-Methylxanthine, For HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.