missing translation for 'onlineSavingsMsg'
Learn More

3-Methylxanthine, MedChemExpress

Product Code. 16407866
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16407866 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16407866 Supplier Medchem Express Supplier No. HY50723

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress 3-Methylxanthine, a xanthine derivative, is a cyclic guanosine monophosphate (GMP) inhibitor, with an IC50 of 920 μM on guinea-pig isolated trachealis muscle.

3-Methylxanthine, a xanthine derivative, is a cyclic guanosine monophosphate (GMP) inhibitor, with an IC50 of 920 μM on guinea-pig isolated trachealis muscle.

Specifications

Chemical Name or Material 3-Methylxanthine
CAS 1076-22-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C6H6N4O2
Quantity 100 mg
Solubility Information 1M NaOH : 50 mg/mL (300.95 mM; ultrasonic and adjust pH to 11 with 1M NaOH) ∣DMSO : 2.61 mg/mL (15.71 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (insoluble) ∣H2O : < 0.1 mg/mL (insoluble) ∣DMF : < 1 mg/mL (insoluble)
SMILES O=C1N(C)C2=C(N=CN2)C(N1)=O
Molecular Weight (g/mol) 166.14
Formula Weight 166.14
Percent Purity 95.75%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.