Learn More
Metrohm 3 mol/L KCI Electrolyte Solution
Product Code. 13253990
Encompass_Preferred
Change view
Click to view available options
Filling Solution Type:
KCI Electrolyte Solution
Product Code. Filling Solution Type
13253990 KCI Electrolyte Solution
1 options
Product Code. 13253990 Supplier Metrohm Supplier No. 6.2308.020
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
An excellent solution for electrode conditioning.
  • Nonhazardous
  • For use with Ag/AgCl reference systems
  • Sold individually in a 250mL bottle
Specifications
For Use With (Equipment) Ag/AgCl Reference Systems
Filling Solution Type KCI Electrolyte Solution
Packaging 250 mL Bottle