missing translation for 'onlineSavingsMsg'
Learn More

3-Morpholino-1-(4-(2-oxopiperidin-1-yl)phenyl)-5,6-dihydropyridin-2(1H)-one, TRC

Product Code. 30137318
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137318 10 g 10g
17920755 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137318 Supplier Toronto Research Chemicals Supplier No. TRCM73278810G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 545445-44-1
Molecular Formula C20H25O3N3
Molecular Weight (g/mol) 355.43
Synonym Apixaban Intermediate 1
IUPAC Name 5-morpholin-4-yl-1-[4-(2-oxopiperidin-1-yl)phenyl]-2,3-dihydropyridin-6-one
SMILES C1CCN(C2=CC=C(C=C2)N3CCC=C(C3=O)N4CCOCC4)C(=O)C1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C20H25N3O3/c24-19-5-1-2-10-22(19)16-6-8-17(9-7-16)23-11-3-4-18(20(23)25)21-12-14-26-15-13-21/h4,6-9H,1-3,5,10-15H2
Formula Weight 355.1896
Percent Purity >95%
Chemical Name or Material 3-Morpholino-1-(4-(2-oxopiperidin-1-yl)phenyl)-5,6-dihydropyridin-2(1H)-one
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.