Learn More
Eppendorf™ Research 3 Neo Adjustment Tool
Product Code. 30689609 Shop All Eppendorf Products
Change view
Click to view available options
Type:
Pipette Adjustment Tool
Unit Size:
Pack of 5
Product Code. Type Unit Size
30689609 Pipette Adjustment Tool Pack of 5
1 options
Product Code. 30689609 Supplier Eppendorf™ Supplier No. 3102690000
  • ONLINE EXCLUSIVE PRICE
    10.90 €
    10.20 € / Pack of 5
    Save 0.70 €
    6% Off
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use the adjustment tool to change factory or temporary settings in Eppendorf Research 3 neo pipettes.
  • Adjusts factory or temporary settings on Eppendorf Research™ plus and 3 neo pipettes
  • Essential for precise calibration and performance optimization
  • Original Eppendorf spare part ensures compatibility and reliability
  • Ideal replacement if the tool from original packaging is misplaced
  • Includes 3 Neo Adjustment Tools per set
TRUSTED_SUSTAINABILITY
Specifications
For Use With Pipettes
Material Polypropylene
Type Pipette Adjustment Tool
For Use With (Equipment) 3 Neo
Quantity 1 Pk.