Learn More
Eppendorf™ Research 3 Neo Factory Adjustment Seal
Product Code. 30689666 Shop All Eppendorf Products
Change view
Click to view available options
Type:
Protective Seal
Unit Size:
Pack of 10
Product Code. Type Unit Size
30689666 Protective Seal Pack of 10
1 options
Product Code. 30689666 Supplier Eppendorf™ Supplier No. 3102603001
  • ONLINE EXCLUSIVE PRICE
    16.25 €
    15.20 € / Pack of 10
    Save 1.05 €
    6% Off
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Factory adjustment seal for Eppendorf Research™ 3 neo
  • Factory Adjustment Seal specifically designed for Eppendorf Research™ 3 Neo pipettes
  • Constructed from high-quality Polypropylene for durability and chemical resistance
  • Acts as a protective seal during factory calibration or pipette adjustments
  • Ensures tamper-evidence and maintains instrument integrity
  • Ideal for maintaining accuracy and compliance in laboratory environments
TRUSTED_SUSTAINABILITY
Specifications
For Use With Pipettes
Material Polypropylene
Type Protective Seal
For Use With (Equipment) 3 Neo, Adjustment Seal
Quantity 1 Pk.