missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Research 3 Neo Pipette Carousel and Holder

Product Code. 30689699 Shop All Eppendorf Products
Click to view available options
Product Type:
Pipette Carousel
Pipette Holder
Positions:
1
6
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30689699

Brand: Eppendorf™ 3116000295

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Our flexible carousels, stands and wall mount devices fit all current Eppendorf manual liquid handling instruments

  • The Eppendorf Pipette Holder System stores all current mechanical and electronic Eppendorf pipettes and most of their predecessors
  • Pipette Carousel 2, white Rotatable holder with 6 holders for Eppendorf Research™ 3 neo pipettes
  • Carousels and Stands with interchangeable pipette holders and charger shells, no tools required
  • Large rubber feet protect carousels and stands from liquids spilled on bench top\n
  • Pictograms on all charger shells and pipette holders for clear assignment in the lab
  • Save valuable bench space with the Eppendorf Pipette Holder System
  • With space for up to six single- or multi-channel pipettes, you can easily store all of your Eppendorf pipettes and multi-dispensers within easy reach
  • Our flexible carousels, stands and wall mount devices fit all current Eppendorf manual liquid handling instruments as well as most of their predecessors
TRUSTED_SUSTAINABILITY

Specifications

Product Type Pipette Holder
Autoclavable Not Autoclavable
Color White
Holds Manual Pipette
Material Polycarbonate
Positions 1
Shape Rectangular
Shelves None
Stackable No
Sterility Non-sterile
For Use With (Equipment) 3 Neo
Quantity 1 Ea.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.