Learn More
Eppendorf™ Research 3 Neo Single Channel Pipettes Locking Ring
Product Code. 30689631 Shop All Eppendorf Products
Change view
Click to view available options
Type:
Locking Clip
Unit Size:
Pack of 5
Product Code. Type Unit Size
30689631 Locking Clip Pack of 5
1 options
Product Code. 30689631 Supplier Eppendorf™ Supplier No. 3102637000
  • ONLINE EXCLUSIVE PRICE
    11.26 €
    10.20 € / Pack of 5
    Save 1.06 €
    9% Off
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use to deactivate the spring-loaded tip cone feature in Eppendorf single-channel pipettes
  • Designed for Eppendorf single-channel pipettes
  • Ideal when using third-party pipette tips
  • Helps deactivate the spring-loaded tip cone feature
  • Ensures secure tip attachment and accurate pipetting
  • Includes 3 Neo Single Channel Pipettes Locking Rings
TRUSTED_SUSTAINABILITY
Specifications
For Use With Pipettes
Material Polypropylene
Type Locking Clip
For Use With (Equipment) Research plus, Reference 2 Single - Channel Pipettes
Quantity 1 Pk.