Learn More
Eppendorf™ Research 3 Neo Spring Lock
Product Code. 30689662 Shop All Eppendorf Products
Click to view available options
Type:
Spring Lock
Unit Size:
Pack of 5
Product Code. 30689662
Brand: Eppendorf™ 3102632000
The spring lock secures the pipette's spring, ideal for replacing worn or damaged spring locks to restore full functionality
  • 3 Neo Spring Lock designed for secure retention of the pipette's internal spring
  • Original Eppendorf spare part ensures compatibility with Research™ 3 Neo pipettes
  • Ideal for replacing worn or damaged spring locks to restore full functionality
  • Maintains pipette performance and reliability during repetitive use
  • Durable construction supports long-term laboratory operation and precision
TRUSTED_SUSTAINABILITY
Specifications
For Use With Pipettes
Material Plastic
Type Spring Lock
Volume (Metric) 2 to 10 mL
For Use With (Equipment) 3 Neo, 2 to 10 mL and MultitoChannel
Quantity 5 Pk.