missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-1,2,4-triazole, 96%

Product Code. 11337016
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11337016 5 g 5g
11327016 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337016 Supplier Thermo Scientific Alfa Aesar Supplier No. L06845.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 24807-55-4 | C2H2N4O2 | 114.064 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24807-55-4
Molecular Formula C2H2N4O2
Molecular Weight (g/mol) 114.064
MDL Number MFCD00009749
InChI Key KUEFXPHXHHANKS-UHFFFAOYSA-N
Synonym 3-nitro-1,2,4-triazole, 3-nitro-1h-1,2,4-triazole, 3-nitro-4h-1,2,4-triazole, 1h-1,2,4-triazole, 3-nitro, s-triazole, 3-nitro, 5-nitro-1h-1,2,4 triazole, 3-nitro-1,2,4 triazole, zlchem 554, 3-nitro-1,4-triazole
PubChem CID 90614
IUPAC Name 5-nitro-1H-1,2,4-triazole
SMILES C1=NNC(=N1)[N+](=O)[O-]

Specifications

Melting Point ∽215°C (decomposition)
Quantity 5 g
Beilstein 607167
Formula Weight 114.06
Percent Purity 96%
Chemical Name or Material 3-Nitro-1,2,4-triazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.