missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-1-pyrenol, TRC

Product Code. 30474056
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474056 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30474056 Supplier Toronto Research Chemicals Supplier No. TRCN51998025MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Nitro-1-pyrenol
CAS 86674-49-9
Molecular Formula C16H9NO3
Quantity 25 mg
Synonym 1-Hydroxy-3-nitropyrene, 1-Nitro-3-hydroxypyrene, 1-Nitropyren-3-ol, 3-Hydroxy-1-nitropyrene
InChI Formula InChI=1S/C16H9NO3/c18-14-8-13(17(19)20)11-6-4-9-2-1-3-10-5-7-12(14)16(11)15(9)10/h1-8,18H
SMILES Oc1cc(c2ccc3cccc4ccc1c2c34)[N+](=O)[O-]
IUPAC Name 3-nitropyren-1-ol
Molecular Weight (g/mol) 263.25
Formula Weight 263.0582
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.