missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-1H-pyrazole, 97%, Thermo Scientific Chemicals

Product Code. 15284884 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
Product Code. Quantity Packaging unitSize
15284884 5 g Glass bottle 5g
15294884 25 g Glass bottle 25g
2 options
This item is not returnable. View return policy

Product Code. 15284884

Brand: Thermo Scientific Acros 451680050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26621-44-3
Molecular Formula C3H3N3O2
Molecular Weight (g/mol) 113.08
MDL Number MFCD00159621,MFCD00238787
InChI Key MZRUFMBFIKGOAL-UHFFFAOYSA-N
Synonym 3-nitro-1h-pyrazole, 3-nitropyrazole, 1h-pyrazole, 3-nitro, 3-nitro-2h-pyrazole, nitropyrazole, pyrazole, 3-nitro, 5-nitro-1h-pyrazol, djw 94, djw-94, 5-nitropyrazole
PubChem CID 123419
IUPAC Name 5-nitro-1H-pyrazole
SMILES [O-][N+](=O)C1=CC=NN1

Specifications

Melting Point 173.0°C to 175.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 113.08
Percent Purity 97%
Chemical Name or Material 3-Nitro-1H-pyrazole
compliance-icons
Product Identifier
  • 5-Nitro-1H-pyrazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.