missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-alpha-toluenesulfonyl chloride, 95%

Product Code. 11490947
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490947 25 g 25g
11470947 1 g 1g
11480947 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490947 Supplier Thermo Scientific Alfa Aesar Supplier No. B23333.14
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 58032-84-1 | C7H6ClNO4S | 235.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Reacts with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58032-84-1
Molecular Formula C7H6ClNO4S
Molecular Weight (g/mol) 235.64
MDL Number MFCD01859935
InChI Key IUYFKLDRPZLAQQ-UHFFFAOYSA-N
Synonym 3-nitrophenyl methanesulfonyl chloride, 3-nitrobenzylsulfonyl chloride, 3-nitrophenylmethanesulfonylchloride, benzenemethanesulfonyl chloride, 3-nitro, 3-nitro-alpha-toluenesulfonyl chloride, chloro 3-nitrophenyl methyl sulfone, acmc-1b04d, 3-nitro-phenylmethane-sulfonyl chloride, 3-nitrophenyl methanesulphonyl chloride, 3-nitrobenzenemethanesulfonyl chloride
PubChem CID 13196775
IUPAC Name (3-nitrophenyl)methanesulfonyl chloride
SMILES [O-][N+](=O)C1=CC=CC(CS(Cl)(=O)=O)=C1

Specifications

Melting Point 95°C to 102°C
Quantity 25 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 235.64
Percent Purity 95%
Chemical Name or Material 3-Nitro-alpha-toluenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.