missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-o-phenylenediamine, 98%

Product Code. 15473207
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
15473207 1 g 1g
15483207 5 g 5g
15493207 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 15473207

Brand: Thermo Scientific Alfa Aesar B20201.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3694-52-8 | C6H7N3O2 | 153.141 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3694-52-8
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.141
MDL Number MFCD00007722
InChI Key IOCXBXZBNOYTLQ-UHFFFAOYSA-N
Synonym 3-nitro-1,2-phenylenediamine, 3-nitro-o-phenylenediamine, 1,2-diamino-3-nitrobenzene, 3-nitrophenylenediamine, 2-amino-6-nitroaniline, 1,2-benzenediamine, 3-nitro, 3-nitro-1,2-benzenediamine, o-phenylenediamine, 3-nitro, unii-wrz63l50zk, ccris 5427
PubChem CID 4359525
IUPAC Name 3-nitrobenzene-1,2-diamine
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])N)N

Specifications

Melting Point 157°C to 162°C
Quantity 1 g
Beilstein 909344
Formula Weight 153.14
Percent Purity 98%
Chemical Name or Material 3-Nitro-o-phenylenediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.