missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-o-phenylenediamine, 98%

Product Code. 10205821 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10205821 5 g Glass bottle 5g
10061771 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10205821 Supplier Thermo Scientific Acros Supplier No. 330850050
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3694-52-8 | C6H7N3O2 | 153.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3694-52-8
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007722
InChI Key IOCXBXZBNOYTLQ-UHFFFAOYSA-N
Synonym 3-nitro-1,2-phenylenediamine, 3-nitro-o-phenylenediamine, 1,2-diamino-3-nitrobenzene, 3-nitrophenylenediamine, 2-amino-6-nitroaniline, 1,2-benzenediamine, 3-nitro, 3-nitro-1,2-benzenediamine, o-phenylenediamine, 3-nitro, unii-wrz63l50zk, ccris 5427
PubChem CID 4359525
IUPAC Name 3-nitrobenzene-1,2-diamine
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])N)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 157.0°C to 161.0°C
Color Red
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 13, I, 10
Formula Weight 153.14
Percent Purity 98%
Physical Form Needle-like Crystals
Chemical Name or Material 3-Nitro-o-phenylenediamine, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.