missing translation for 'onlineSavingsMsg'
Learn More

3-Nitroaniline, 98%

Product Code. 10635372 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10635372 500 g Glass bottle 500g
10459630 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10635372 Supplier Thermo Scientific Acros Supplier No. 128365000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-09-2 | C6H6N2O2 | 138.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-09-2
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00007782
InChI Key XJCVRTZCHMZPBD-UHFFFAOYSA-N
Synonym m-nitroaniline, benzenamine, 3-nitro, 3-nitrophenylamine, nitranilin, 3-aminonitrobenzene, fast orange base r, azobase mna, devol orange r, m-nitrophenylamine, 3-nitrobenzenamine
PubChem CID 7423
IUPAC Name 3-nitroaniline
SMILES NC1=CC=CC(=C1)[N+]([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 111.0°C to 114.0°C
Color Orange to Yellow
Boiling Point 306.0°C
Flash Point 196°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4NH2
Quantity 500 g
Beilstein 12, 698
Merck Index 15, 6668
Solubility Information Solubility in water: 1.25g/L in water. Other solubilities: soluble in alcohol,ether,methanol and ethanol,1 g soluble in 20 mL alcohol,1 g soluble in 18 mL ether,1 g soluble in 11.5 mL methanol
Formula Weight 138.13
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 3-Nitroaniline, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Nitroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.