missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzaldoxime, 98%, Thermo Scientific™

Product Code. 11327256
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327256 100 g 100g
11317256 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327256 Supplier Thermo Scientific Alfa Aesar Supplier No. L09144.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3431-62-7
Molecular Formula C7H6N2O3
Molecular Weight (g/mol) 166.136
MDL Number MFCD00007275
InChI Key GQMMRLBWXCGBEV-YVMONPNESA-N
Synonym 1z-3-nitrobenzaldehyde oxime, m-nitrobenzaldehyde oxime, 3-nitrobenzaldehyde oxime, nz-n-3-nitrophenyl methylidene hydroxylamine, m-nitrobenzaldoxime, anti-m-nitrobenzaldoxime, z-m-nitrobenzaldoxime, 3-nitrobenzaldoxime, nsc34, e-3-nitrobenzaldehyde oxime
PubChem CID 5369356
IUPAC Name (NZ)-N-[(3-nitrophenyl)methylidene]hydroxylamine
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C=NO

Specifications

Melting Point 122°C to 125°C
Quantity 100 g
UN Number UN2811
Beilstein 2047081
Formula Weight 166.14
Percent Purity 98%
Chemical Name or Material 3-Nitrobenzaldoxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.