missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzenesulfonyl chloride, 97%

Product Code. 10781442 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10781442 25 g Glass bottle 25g
10117800 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10781442 Supplier Thermo Scientific Acros Supplier No. 415840250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 121-51-7 | C6H4ClNO4S | 221.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-51-7
Molecular Formula C6H4ClNO4S
Molecular Weight (g/mol) 221.62
MDL Number MFCD00007435
InChI Key MWWNNNAOGWPTQY-UHFFFAOYSA-N
Synonym 3-nitrophenylsulfonyl chloride, 3-nitrobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 3-nitro, 3-nitrobenzenesulphonyl chloride, 3-nitro-benzenesulfonyl chloride, m-nitrophenylsulfonyl chloride, m-nitrobenzenesulfonyl chloride, unii-kn1z8wpl9u, benzenesulfonyl chloride, m-nitro, kn1z8wpl9u
PubChem CID 8476
IUPAC Name 3-nitrobenzenesulfonyl chloride
SMILES C1=CC(=CC(=C1)S(=O)(=O)Cl)[N+](=O)[O-]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 60.0°C to 65.0°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula SO2ClC6H4NO2
Quantity 25 g
Solubility Information Solubility in water: decomposes
Formula Weight 221.62
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrobenzenesulfonyl chloride, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.