missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzenesulfonyl chloride, 98%

Product Code. 11463248
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463248 5 g 5g
11483248 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11463248 Supplier Thermo Scientific Alfa Aesar Supplier No. A17670.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 121-51-7 | C6H4ClNO4S | 221.611 g/mol

It is employed as a biochemical for proteomics research. It is also used in the preparation of: acyl-2-aminobenzimidazole analogs and 6-chloro-2-methyl-3-{1-[(3-nitrophenyl)sulfonyl]-1H-pyrazol-3-yl}imidazo[1,2-a]pyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a biochemical for proteomics research. It is also used in the preparation of: acyl-2-aminobenzimidazole analogs and 6-chloro-2-methyl-3-{1-[(3-nitrophenyl)sulfonyl]-1H-pyrazol-3-yl}imidazo[1,2-a]pyridine.

Solubility
Hydrolyzes slowly in water.

Notes
Moisture Sensitive. Store away from oxidizing agents, active metals and bases. Keep the container tightly closed in a cool, dry place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-51-7
Molecular Formula C6H4ClNO4S
Molecular Weight (g/mol) 221.611
MDL Number MFCD00007435
InChI Key MWWNNNAOGWPTQY-UHFFFAOYSA-N
Synonym 3-nitrophenylsulfonyl chloride, 3-nitrobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 3-nitro, 3-nitrobenzenesulphonyl chloride, 3-nitro-benzenesulfonyl chloride, m-nitrophenylsulfonyl chloride, m-nitrobenzenesulfonyl chloride, unii-kn1z8wpl9u, benzenesulfonyl chloride, m-nitro, kn1z8wpl9u
PubChem CID 8476
IUPAC Name 3-nitrobenzenesulfonyl chloride
SMILES C1=CC(=CC(=C1)S(=O)(=O)Cl)[N+](=O)[O-]

Specifications

Melting Point 59°C to 63°C
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3261
Beilstein 746542
Formula Weight 221.62
Percent Purity 98%
Chemical Name or Material 3-Nitrobenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.