missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzhydrazide, 98+%

Product Code. 11495706
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495706 5 g 5g
11405716 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495706 Supplier Thermo Scientific Alfa Aesar Supplier No. B20800.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 618-94-0 | C7H7N3O3 | 181.151 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-94-0
Molecular Formula C7H7N3O3
Molecular Weight (g/mol) 181.151
MDL Number MFCD00017059
InChI Key NQEWXLVDAVTOHM-UHFFFAOYSA-N
Synonym 3-nitrobenzhydrazide, 3-nitrobenzoylhydrazine, m-nitrobenzhydrazide, 3-nitrobenzoic acid hydrazide, benzoic acid, 3-nitro-, hydrazide, m-nitrobenzoylhydrazide, m-nitrobenzoylhydrazine, m-nitrobenzoic hydrazide, m-nitrobenzoic acid hydrazide, benzoic acid, m-nitro-, hydrazide
PubChem CID 3291498
IUPAC Name 3-nitrobenzohydrazide
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)NN

Specifications

Melting Point 152°C to 155°C
Quantity 5 g
Beilstein 645512
Formula Weight 181.15
Percent Purity ≥98%
Chemical Name or Material 3-Nitrobenzhydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.