missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzonitrile, 98%

Product Code. 10645232 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10645232 25 g Glass bottle 25g
10580784 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10645232 Supplier Thermo Scientific Acros Supplier No. 173520250
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 619-24-9 | C7H4N2O2 | 148.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-24-9
Molecular Formula C7H4N2O2
Molecular Weight (g/mol) 148.12
MDL Number MFCD00007194
InChI Key RUSAWEHOGCWOPG-UHFFFAOYSA-N
Synonym benzonitrile, 3-nitro, m-nitrobenzonitrile, 3-cyanonitrobenzene, m-cyanonitrobenzene, benzonitrile, m-nitro, 3-cyano-1-nitrobenzene, 3-nitro-benzonitrile, ccris 2327, 3-nitrobenzenecarbonitrile, 3-nitrobenzonitril
PubChem CID 12079
IUPAC Name 3-nitrobenzonitrile
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 115°C to 119°C
Color Yellow
Boiling Point 165°C (21.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula O2NC6H4CN
Quantity 25 g
Beilstein 09, 385
Solubility Information Solubility in water: insoluble. Other solubilities: freele soluble in ether and alcohol
Formula Weight 148.12
Percent Purity 98%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 3-Nitrobenzonitrile, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Nitrobenzonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.