missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzoyl chloride, 98%

Product Code. 10512751 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10512751 100 g Glass bottle 100g
10327810 5 g Glass bottle 5g
10153490 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10512751 Supplier Thermo Scientific Acros Supplier No. 155841000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 121-90-4 | C7H4ClNO3 | 185.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-90-4
Molecular Formula C7H4ClNO3
Molecular Weight (g/mol) 185.57
MDL Number MFCD00007247
InChI Key NXTNASSYJUXJDV-UHFFFAOYSA-N
Synonym m-nitrobenzoyl chloride, benzoyl chloride, 3-nitro, benzoyl chloride, m-nitro, 3-nitro-benzoyl chloride, ccris 1186, m-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove, 3-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove czech, m-nitrobenzoylchlorid
PubChem CID 8495
IUPAC Name 3-nitrobenzoyl chloride
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 30.0°C to 35.0°C
Color Brown
Density 1.4200g/mL
Boiling Point 275.0°C to 278.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula O2NC6H4COCl
Quantity 100 g
Beilstein 09, 381
Specific Gravity 1.42
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in ether,decomposes in alcohol
Formula Weight 185.57
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material 3-Nitrobenzoyl chloride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.