missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophthalic anhydride, 97%

Product Code. 11472548
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472548 25 g 25g
11482548 100 g 100g
11492548 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472548 Supplier Thermo Scientific Alfa Aesar Supplier No. A15857.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 641-70-3 | C8H3NO5 | 193.11 g/mol

4-Nitrophthalic Anhydride is an intermediate for the synthesis of the benzimidazole PARP inhibitor I, ABT-472. It reacts with aminoquinazolinones to yield phthalimidoquinazolinones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophthalic Anhydride is an intermediate for the synthesis of the benzimidazole PARP inhibitor I, ABT-472. It reacts with aminoquinazolinones to yield phthalimidoquinazolinones.

Solubility
May decompose in water.

Notes
Air sensitive. Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 641-70-3
Molecular Formula C8H3NO5
Molecular Weight (g/mol) 193.11
MDL Number MFCD00005921
InChI Key ROFZMKDROVBLNY-UHFFFAOYSA-N
Synonym 3-nitrophthalic anhydride, 4-nitroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 4-nitro, phthalic anhydride, 3-nitro, 3-nitro phthalic anhydride, 3-nitrophthalic acid anhydride, 1,3-isobenzofurandione, nitro, 4-nitro-isobenzofuran-1,3-dione, 3-nitrophthalicanhydride, nitrophthalic anhydride
PubChem CID 21631
SMILES [O-][N+](=O)C1=CC=CC2=C1C(=O)OC2=O

Specifications

Melting Point 161°C to 166°C
Quantity 25 g
Beilstein 179963
Sensitivity Moisture sensitive
Solubility Information May decompose in water.
IUPAC Name 4-nitro-1,3-dihydro-2-benzofuran-1,3-dione
Formula Weight 193.11
Percent Purity 97%
Chemical Name or Material 3-Nitrophthalic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.