missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophthalic anhydride, 97%

Product Code. 10645032 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10645032 25 g Glass bottle 25g
10285690 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10645032 Supplier Thermo Scientific Acros Supplier No. 168990250
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 641-70-3 | C8H3NO5 | 193.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 641-70-3
Molecular Formula C8H3NO5
Molecular Weight (g/mol) 193.11
MDL Number MFCD00005921
InChI Key ROFZMKDROVBLNY-UHFFFAOYSA-N
Synonym 3-nitrophthalic anhydride, 4-nitroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 4-nitro, phthalic anhydride, 3-nitro, 3-nitro phthalic anhydride, 3-nitrophthalic acid anhydride, 1,3-isobenzofurandione, nitro, 4-nitro-isobenzofuran-1,3-dione, 3-nitrophthalicanhydride, nitrophthalic anhydride
PubChem CID 21631
IUPAC Name 4-nitro-2-benzofuran-1,3-dione
SMILES [O-][N+](=O)C1=CC=CC2=C1C(=O)OC2=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 160.0°C to 165.0°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Beilstein 17, 486
Solubility Information Solubility in water: decomposes.
Formula Weight 193.12
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrophthalic anhydride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Nitrophthalic anhydride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.