missing translation for 'onlineSavingsMsg'
Learn More

3-Nonanol, 98%

Product Code. 11469416
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469416 250 g 250g
11459416 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469416 Supplier Thermo Scientific Alfa Aesar Supplier No. A19120.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 624-51-1 | C9H20O | 144.258 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-51-1
Molecular Formula C9H20O
Molecular Weight (g/mol) 144.258
MDL Number MFCD00014411
InChI Key GYSCXPVAKHVAAY-UHFFFAOYSA-N
Synonym 3-nonanol, hexylethylcarbinol, ethylhexylcarbinol, 3-nonyl alcohol, nonan-7-ol, ethyl hexyl carbinol, acmc-1b2h7
PubChem CID 12216
IUPAC Name nonan-3-ol
SMILES CCCCCCC(CC)O

Specifications

Density 0.824
Boiling Point 192°C to 194°C
Flash Point 79°C (174°F)
Refractive Index 1.431
Quantity 250 g
Beilstein 1719453
Formula Weight 144.26
Percent Purity 98%
Chemical Name or Material 3-Nonanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.