missing translation for 'onlineSavingsMsg'
Learn More

3-O-Methyl-D-Glucose Crystalline powder MP Biomedicals

Product Code. 11482372 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482372 25 g 25g
11462372 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11482372 Supplier MP Biomedicals Supplier No. 0215165125
  • ONLINE EXCLUSIVE PRICE
    959.00 €
    909.30 € / 25g
    Save 49.70 €
    5% Off
 Restricted Item
Estimated Shipment: 23-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 13224-94-7
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00063266
InChI Key SCBBSJMAPKXHAH-OVHBTUCOSA-N
Synonym 3-o-methyl-alpha-d-glucopyranose, 3-o-methyl-a-d-glucopyranose, 2s,3r,4s,5r,6r-6-hydroxymethyl-4-methoxytetrahydro-2h-pyran-2,3,5-triol, 2s,3r,4s,5r,6r-6-hydroxymethyl-4-methoxyoxane-2,3,5-triol, 3-o-methyl-, a-d-glucopyranose, 3-o-methyl-alpha-d-glucose, a-d-glucopyranose,3-o-methyl, alpha-d-glucopyranose, 3-o-methyl, 3-o-methyl-d-glucopyranose
PubChem CID 83246
IUPAC Name (2S,3R,4S,5R,6R)-6-(hydroxymethyl)-4-methoxyoxane-2,3,5-triol
SMILES CO[C@@H]1[C@@H](O)[C@@H](O)O[C@H](CO)[C@H]1O

Specifications

Color White
Quantity 25 g
Formula Weight 194.2
Physical Form Powder
Chemical Name or Material 3-O-methyl-D-glucose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.