missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3'-O-Sulfate-Epicatechin Ammonium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 3'-O-Sulfate-Epicatechin Ammonium Salt |
| CAS | 1421313-26-9 |
| Molecular Formula | C15 H14 O9 S . H3 N |
| Quantity | 10 mg |
| Synonym | (2R,3R)-3,4-Dihydro-2-[4-hydroxy-3-(sulfooxy)phenyl]-2H-1-benzopyran-3,5,7-triol Ammonium Salt |
| InChI Formula | InChI=1S/C15H14O9S.H3N/c16-8-4-11(18)9-6-12(19)15(23-13(9)5-8)7-1-2-10(17)14(3-7)24-25(20,21)22;/h1-5,12,15-19H,6H2,(H,20,21,22);1H3/t12-,15-;/m1./s1 |
| SMILES | N.O[C@@H]1Cc2c(O)cc(O)cc2O[C@@H]1c3ccc(O)c(OS(=O)(=O)O)c3 |
| IUPAC Name | azane;[2-hydroxy-5-[(2R,3R)-3,5,7-trihydroxy-3,4-dihydro-2H-chromen-2-yl]phenyl] hydrogen sulfate |
| Molecular Weight (g/mol) | 387.362 |
| Formula Weight | 387.062 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?