missing translation for 'onlineSavingsMsg'
Learn More

3-Oxetanone, 95%

Product Code. 15484258
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15484258 1 g 1g
15494258 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15484258 Supplier Thermo Scientific Alfa Aesar Supplier No. H57870.03
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 6704-31-0 | C3H4O2 | 72.063 g/mol

3-Oxetanone is used as a starting material for the preparation of other oxetanes compounds, which are of pharmacological interest. It is also considered as an object for theoretical studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Oxetanone is used as a starting material for the preparation of other oxetanes compounds, which are of pharmacological interest. It is also considered as an object for theoretical studies.

Solubility
Miscible with tetrahydrofuran.

Notes
Incompatible with strong oxidizing agents. Heat sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6704-31-0
Molecular Formula C3H4O2
Molecular Weight (g/mol) 72.063
MDL Number MFCD09263255
InChI Key ROADCYAOHVSOLQ-UHFFFAOYSA-N
Synonym 3-oxetanone, 1,3-epoxy-2-propanone, 3-oxooxetane, oxetane-3-one, oxetan-3-on, 1,3-epoxypropanone, pubchem15880, 3-oxentanone, acmc-209nxj, ksc358e8n
PubChem CID 15024254
IUPAC Name oxetan-3-one
SMILES C1C(=O)CO1

Specifications

Density 1.124
Boiling Point 140°C
Flash Point 26°C (79°F)
Refractive Index 1.426
Quantity 1 g
UN Number UN1993
Solubility Information Miscible with tetrahydrofuran.
Formula Weight 72.06
Percent Purity 95%
Chemical Name or Material 3-Oxetanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.